Methyl-4-aminobicyclo[2.2.2]octane-1-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-aminobicyclo[2.2.2]octane-1-carboxylate hydrochloride, 4-Aminobicyclo[2.2.2]octane-1-carboxylic acid methyl ester
OR312511-250MG
APOSOR312511-250MGEA
75.4
EUR
APOSOR312511-250MG
Methyl-4-aminobicyclo[2.2.2]octane-1-carboxylate
Methyl-4-aminobicyclo[2.2.2]octane-1-carboxylate
Formel:
C₁₀H₁₇NO₂ Molecular Weight: 183,2475 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20693756 CAS-Nummer: 135908-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...