1-(1H-Imidazol-2-yl)ethanone 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Acetylimidazole
OR313011-1G
OR313011-5G
APOSOR313011-1GEA
101
EUR
APOSOR313011-1G
APOSOR313011-5G
1-(1H-Imidazol-2-yl)ethanone 97%
1-(1H-Imidazol-2-yl)ethanone
Formel:
C₅H₆N₂O Molecular Weight: 110,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878742 CAS-Nummer: 53981-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...