Dimethyl (1-diazo-2-oxopropyl)phosphonate 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Ohira-Bestmann Reagent, 1-Diazoacetonylphosphonic Acid Dimethyl Ester, Dimethyl 1-Diazoacetonylphosphonate, (1-Diazo-2-oxopropyl)phosphonic Acid Dimethyl Ester
OR313045-5G
OR313045-25G
OR313045-100G
APOSOR313045-5GEA
94.4
EUR
APOSOR313045-5G
APOSOR313045-25G
APOSOR313045-100G
Dimethyl (1-diazo-2-oxopropyl)phosphonate 95%
Dimethyl (1-diazo-2-oxopropyl)phosphonate
Formel:
C₅H₉N₂O₄P Molecular Weight: 192,11 g/mol Dichte: 1,28 g/cm³ (25 °C) Flammpunkt: 2 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07368360 CAS-Nummer: 90965-06-3 UN: 1648 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...