3,3-Dimethylazetidine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR313047-250MG OR313047-1G OR313047-5G
APOSOR313047-250MGEA 78.3 EUR
APOSOR313047-250MG APOSOR313047-1G APOSOR313047-5G
3,3-Dimethylazetidine hydrochloride
3,3-Dimethylazetidine hydrochloride
Formel: C₅H₁₂ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD10565785
CAS-Nummer: 89381-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR