2-Chlor-5-hydroxybenzaldehyd
Lieferant: Apollo Scientific
OR313096-1G
OR313096-5G
OR313096-25G
APOSOR313096-1GEA
55.1
EUR
APOSOR313096-1G
APOSOR313096-5G
APOSOR313096-25G
2-Chlor-5-hydroxybenzaldehyd
2-Chloro-5-hydroxybenzaldehyde
Formel:
C₇H₅ClO₂ Molecular Weight: 156,57 g/mol Schmelzpunkt: 110,5…111.5 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07783652 CAS-Nummer: 7310-94-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...