2-Hydroxypyridine-4-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Hydroxyisonicotinaldehyde, 2-Hydroxy-4-formylpyridine
OR313101-1G
OR313101-250MG
APOSOR313101-1GEA
140
EUR
APOSOR313101-1G
APOSOR313101-250MG
2-Hydroxypyridine-4-carbaldehyde
2-Hydroxypyridine-4-carbaldehyde
Formel:
C₆H₅NO₂ Molecular Weight: 123,11 g/mol Schmelzpunkt: 79…81 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03788455 CAS-Nummer: 188554-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...