tert-Butyl-4-brombenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 4-bromobenzoate

OR313102-25G
APOSOR313102-25GEA 108 EUR
APOSOR313102-25G
tert-Butyl-4-brombenzoat
tert-Butyl-4-brombenzoat
Formel: C₁₁H₁₃BrO₂
Molecular Weight: 257,1200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00093266
CAS-Nummer: 59247-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR