2-Amino-3-benzyloxypyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Benzyloxypyridin-2-amin, 3-Benzyloxy-2-pyridylamin
OR3137-25G
OR3137-5G
APOSOR3137-25GEA
40.6
EUR
APOSOR3137-25G
APOSOR3137-5G
2-Amino-3-benzyloxypyridin
2-Amino-3-benzyloxypyridin
Formel:
C₁₂H₁₂N₂O Molecular Weight: 200,24 g/mol Schmelzpunkt: 92…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006316 CAS-Nummer: 24016-03-3 EINECS: 245-983-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...