3-Amino-2-pyridone 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-2-hydroxypyridine, 3-Amino-2-pyridinol, 3-Amino-2-pyridinone

OR315027-1G OR315027-25G OR315027-5G
APOSOR315027-1GEA 50.9 EUR
APOSOR315027-1G APOSOR315027-25G APOSOR315027-5G
3-Amino-2-pyridone 98%
3-Amino-2-pyridone
Formel: C₅H₆N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD03411556
CAS-Nummer: 33630-99-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR