Isochinolin-1-amin

Lieferant: Apollo Scientific

Synonyme: Isoquinolin-1-amine, 1-Aminoisoquinoline

OR315041-5G
APOSOR315041-5GEA 68.3 EUR
APOSOR315041-5G
Isochinolin-1-amin
Isochinolin-1-amin
Formel: C₉H₈N₂
Molecular Weight: 144,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00024137
CAS-Nummer: 1532-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR