1-Isochinolinol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Isocarbostyril, 1-Hydroxyisoquinoline
OR315042-100G
OR315042-25G
OR315042-5G
APOSOR315042-100GEA
406
EUR
APOSOR315042-100G
APOSOR315042-25G
APOSOR315042-5G
1-Isochinolinol
1-Isochinolinol
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol Schmelzpunkt: 212…215 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006899 CAS-Nummer: 491-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...