2-Bromo-5-chloronicotinic acid 98%
Lieferant: Apollo Scientific
OR315083-1G
OR315083-5G
OR315083-10G
APOSOR315083-1GEA
105
EUR
APOSOR315083-1G
APOSOR315083-5G
APOSOR315083-10G
2-Bromo-5-chloronicotinic acid 98%
2-Bromo-5-chloronicotinic acid
Formel:
C₆H₃BrClNO₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
65550-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...