5-Amino-2,3-dichlorpyridin 98%

Lieferant: Apollo Scientific

Synonyme: 2,3-Dichlorpyridin-5-amin, 2,3-Dichlor-5-pyridylamin

OR315086-25G
APOSOR315086-25GEA 150 EUR
APOSOR315086-25G
5-Amino-2,3-dichlorpyridin 98%
5-Amino-2,3-dichlorpyridin
Formel: C₅H₄Cl₂N₂
Molecular Weight: 163,01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840434
CAS-Nummer: 98121-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR