Methyl-4-methylnicotinate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-methylnicotinate

OR315098-1G OR315098-10G OR315098-5G
APOSOR315098-1GEA 78.3 EUR
APOSOR315098-1G APOSOR315098-10G APOSOR315098-5G
Methyl-4-methylnicotinate 98%
Methyl-4-methylnicotinate
Formel: C₈H₉NO₂
Molecular Weight: 151,1600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258869
CAS-Nummer: 33402-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR