5-Bromo-2-ethylpyridine 98%

Lieferant: Apollo Scientific
Achtung

OR315121-1G OR315121-5G
APOSOR315121-1GEA 68.3 EUR
APOSOR315121-1G APOSOR315121-5G
5-Bromo-2-ethylpyridine 98%
5-Bromo-2-ethylpyridine
Formel: C₇H₈BrN
Molecular Weight: 186.0491 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226235
CAS-Nummer: 38749-90-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR