4-Bromodibenzofuran 97%

Lieferant: Apollo Scientific

Synonyme: 4-Bromodibenzo[b,d]furan

OR315128-100G
APOSOR315128-100GEA 136 EUR
APOSOR315128-100G
4-Bromodibenzofuran 97%
4-Bromodibenzofuran
Formel: C₁₂H₇BrO
Molecular Weight: 247,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00185685
CAS-Nummer: 89827-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR