5-Brom-3-chlor-2-methoxypyridin 98%
Lieferant: Apollo Scientific
Synonyme:
2-Methoxy-3-chloro-5-bromopyridine
OR315131-5G
APOSOR315131-5GEA
55.1
EUR
APOSOR315131-5G
5-Brom-3-chlor-2-methoxypyridin 98%
5-Brom-3-chlor-2-methoxypyridin
Formel:
C₆H₅BrClNO Molecular Weight: 222,47 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972382 CAS-Nummer: 848366-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...