tert-Butyl bis(2-chloroethyl)carbamate 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: N-BOC-N,N-Bis(2-chloroethyl)amine

OR315132-5G OR315132-100G OR315132-25G
APOSOR315132-5GEA 26.1 EUR
APOSOR315132-5G APOSOR315132-100G APOSOR315132-25G
tert-Butyl bis(2-chloroethyl)carbamate 95%
tert-Butyl bis(2-chloroethyl)carbamate
Formel: C₉H₁₇Cl₂NO₂
Molecular Weight: 242,1428 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06858679
CAS-Nummer: 118753-70-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR