1-Acetyl-7-bromoindolin-5-amine 95+%
Lieferant: Apollo Scientific
OR315195-1G
OR315195-5G
OR315195-10G
APOSOR315195-1GEA
397
EUR
APOSOR315195-1G
APOSOR315195-5G
APOSOR315195-10G
1-Acetyl-7-bromoindolin-5-amine 95+%
1-Acetyl-7-bromoindolin-5-amine
Formel:
C₁₀H₁₁BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09753940 CAS-Nummer: 858193-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...