Methyl-5-amino-2-chlorbenzoat 95+%
Lieferant: Apollo Scientific
Synonyme:
Methyl 5-amino-2-chlorobenzoate
OR315330-1G
OR315330-5G
APOSOR315330-1GEA
21.9
EUR
APOSOR315330-1G
APOSOR315330-5G
Methyl-5-amino-2-chlorbenzoat 95+%
Methyl-5-amino-2-chlorbenzoat
Formel:
C₈H₈ClNO₂ Molecular Weight: 185,6100 g/mol Siedepunkt: 160 °C Schmelzpunkt: 28 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07800315 CAS-Nummer: 42122-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...