1-Boc-4-(aminomethyl)-4-hydroxypiperidine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate, 1-Boc-4-(Aminomethyl)-4-hydroxypiperidine

OR315339-5G
APOSOR315339-5GEA 144 EUR
APOSOR315339-5G
1-Boc-4-(aminomethyl)-4-hydroxypiperidine 95%
1-Boc-4-(aminomethyl)-4-hydroxypiperidine
Formel: C₁₁H₂₂N₂O₃
Molecular Weight: 230,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11579803
CAS-Nummer: 392331-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR