3,6-Dibromcarbazol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,6-Dibromo-9H-carbazole

OR315388-100G OR315388-25G
APOSOR315388-100GEA 45.1 EUR
APOSOR315388-100G APOSOR315388-25G
3,6-Dibromcarbazol 97%
3,6-Dibromocarbazole
Formel: C₁₂H₇Br₂N
Molecular Weight: 325 g/mol
Schmelzpunkt: 204…206 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004961
CAS-Nummer: 6825-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR