Ethyl-1-cyanocyclopropanecarboxylate 96%

Lieferant: Apollo Scientific

Synonyme: Ethyl 1-Cyano-1-cyclopropanecarboxylate

OR315517-100G
APOSOR315517-100GEA 140 EUR
APOSOR315517-100G
Ethyl-1-cyanocyclopropanecarboxylate 96%
Ethyl-1-cyanocyclopropanecarboxylate
Formel: C₇H₉NO₂
Molecular Weight: 139,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00040159
CAS-Nummer: 1558-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR