1-(Diphenylmethyl)-3-azetidinyl-methansulfonat 97%

Lieferant: Apollo Scientific

Synonyme: 1-(Diphenylmethyl)-3-azetidinyl Methanesulphonate, 1-Benzhydrylazetidin-3-yl methanesulphonate, 1-(Diphenylmethyl)-3-(methanesulphonyloxy)azetidine, 1-Benzhydryl-3-(methanesulphonyloxy)azetidine

OR315570-100G
APOSOR315570-100GEA 121 EUR
APOSOR315570-100G
1-(Diphenylmethyl)-3-azetidinyl-methansulfonat 97%
1-Benzhydryl-3-azetidinyl methanesulphonate
Formel: C₁₇H₁₉NO₃S
Molecular Weight: 317,40 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00159216
CAS-Nummer: 33301-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR