9-Hydroxynonanoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR315586-1G
OR315586-250MG
OR315586-5G
APOSOR315586-1GEA
66.7
EUR
APOSOR315586-1G
APOSOR315586-250MG
APOSOR315586-5G
9-Hydroxynonanoic acid 95%
9-Hydroxynonanoic acid
Formel:
C₉H₁₈O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792445 CAS-Nummer: 3788-56-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...