5-Oxopyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific

OR315602-1G OR315602-5G
APOSOR315602-1GEA 132 EUR
APOSOR315602-1G APOSOR315602-5G
5-Oxopyrrolidine-3-carboxylic acid
5-Oxopyrrolidine-3-carboxylic acid
Formel: C₅H₇NO₃
Molecular Weight: 129.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864580
CAS-Nummer: 7268-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR