4'-Phenylacetophenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
p-Phenylacetophenon, 1-(4-Phenylphenyl)ethanon, 4-Phenyl-1-acetylbenzol, 4-Acetylbiphenyl
OR315759-100G
OR315759-1KG
OR315759-500G
APOSOR315759-100GEA
29
EUR
APOSOR315759-100G
APOSOR315759-1KG
APOSOR315759-500G
4'-Phenylacetophenon
4'-Phenylacetophenon
Formel:
C₁₄H₁₂O Molecular Weight: 196.25 g/mol Siedepunkt: 325 Schmelzpunkt: 116…118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008749 CAS-Nummer: 92-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...