10,11-Dihydro-11-oxodibenzo[b,f][1,4]thiazepine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Quetiapine Related Compound G, Dibenzo[b,f][1,4]thiazepin-11(10H)-one

OR315768-100G OR315768-25G OR315768-5G
APOSOR315768-100GEA 121 EUR
APOSOR315768-100G APOSOR315768-25G APOSOR315768-5G
10,11-Dihydro-11-oxodibenzo[b,f][1,4]thiazepine 98%
10,11-Dihydro-11-oxodibenzo[b,f][1,4]thiazepine
Formel: C₁₃H₉NOS
Molecular Weight: 227,2817 g/mol
Schmelzpunkt: 265 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00901197
CAS-Nummer: 3159-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR