Adamantan-1-yl acrylate
Lieferant: Apollo Scientific
Synonyme:
Acrylic acid Adamantan-1-yl ester, 1-Acryloyloxyadamantane
OR315783-5G
APOSOR315783-5GEA
166
EUR
APOSOR315783-5G
Adamantan-1-yl acrylate
Adamantan-1-yl acrylate
Formel:
C₁₃H₁₈O₂ Molecular Weight: 206,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18072700 CAS-Nummer: 121601-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...