(S)-(+)-2-Methoxy-1-phenylethylamine 95%

Lieferant: Apollo Scientific

Synonyme: (S)-(+)-1-Amino-1-phenyl-2-methoxyethane, (S)-2-Methoxy-1-phenylethanamine

OR316003-1G OR316003-250MG
APOSOR316003-1GEA 284 EUR
APOSOR316003-1G APOSOR316003-250MG
(S)-(+)-2-Methoxy-1-phenylethylamine 95%
(S)-(+)-2-Methoxy-1-phenylethylamine
Formel: C₉H₁₃NO
Molecular Weight: 151.2100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01862273
CAS-Nummer: 91298-74-7
UN: 1760
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR