Ethyl-2-oxocyclopentaneacetate
Lieferant: Apollo Scientific
Synonyme: 2-Oxocyclopentaneacetic Acid Ethyl Ester, Ethyl 2-oxocyclopentylacetate, Ethyl 2-(2-Oxocyclopentyl)acetate, 2-(2-Oxocyclopentyl)acetic Acid Ethyl Ester, Ethyl 2-(2-oxocyclopentyl)acetate
OR316028-5G
APOSOR316028-5GEA 82.8 EUR
APOSOR316028-5G
Ethyl-2-oxocyclopentaneacetate
Ethyl-2-oxocyclopentaneacetate
Formel: C₉H₁₄O₃
Molecular Weight: 170,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00044715
CAS-Nummer: 20826-94-2