N-Boc-D-phenylalaninol
Lieferant: Apollo Scientific
Synonyme:
Boc-D-Phenylalaninol
OR316042-100G
OR316042-25G
OR316042-5G
APOSOR316042-100GEA
221
EUR
APOSOR316042-100G
APOSOR316042-25G
APOSOR316042-5G
N-Boc-D-phenylalaninol
N-Boc-D-phenylalaninol
Formel:
C₁₄H₂₁NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216472 CAS-Nummer: 106454-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...