(2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one

Lieferant: Apollo Scientific

Synonyme: (3R,7aS)-3-(Trichloromethyl)tetrahydropyrrolo[1,2-c]oxazol-1(3H)-one, (3R)-Trichloromethyl-cis-Tetrahydropyrrolo[1,2-c]oxazol-1-one, DK7774

OR316050-25G
APOSOR316050-25GEA 98.6 EUR
APOSOR316050-25G
(2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one
(2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one
Formel: C₇H₈Cl₃NO₂
Molecular Weight: 244,50 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08274464
CAS-Nummer: 97538-67-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR