(R)-2-Amino-2-phenylacetamide 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
D(-)-Phenylglycinamide, H-D-Phg-NH₂·HCl, H-D-Phg-NH2
OR316055-100G
OR316055-25G
OR316055-5G
APOSOR316055-100GEA
132
EUR
APOSOR316055-100G
APOSOR316055-25G
APOSOR316055-5G
(R)-2-Amino-2-phenylacetamide 98%
(R)-2-Amino-2-phenylacetamide
Formel:
C₈H₁₀N₂O Molecular Weight: 150,181 g/mol Schmelzpunkt: 125…129 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06799064 CAS-Nummer: 6485-67-2 EINECS: 420-370-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...