tert-Butyl-3-hydroxy-4-methylenepiperidine-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: N-t-BOC-4-Methylene-3-Piperidinol, tert-Butyl 3-hydroxy-4-methylenepiperidine-1-carboxylate

OR316061-1G OR316061-250MG
APOSOR316061-1GEA 162 EUR
APOSOR316061-1G APOSOR316061-250MG
tert-Butyl-3-hydroxy-4-methylenepiperidine-1-carboxylate
tert-Butyl-3-hydroxy-4-methylenepiperidine-1-carboxylate
Formel: C₁₁H₁₉NO₃
Molecular Weight: 213,2735 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15474939
CAS-Nummer: 159635-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR