1-Bromocarbazole 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-9H-carbazole

OR31659-1G OR31659-5G OR31659-25G
APOSOR31659-1GEA 46.4 EUR
APOSOR31659-1G APOSOR31659-5G APOSOR31659-25G
1-Bromocarbazole 99%
1-Bromocarbazole
Formel: C₁₂H₈BrN
Molecular Weight: 246,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18450164
CAS-Nummer: 16807-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR