4-Bromocarbazole 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-9H-carbazole

OR31660-1G OR31660-5G OR31660-25G
APOSOR31660-1GEA 21.9 EUR
APOSOR31660-1G APOSOR31660-5G APOSOR31660-25G
4-Bromocarbazole 99%
4-Bromocarbazole
Formel: C₁₂H₈BrN
Molecular Weight: 246,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18450175
CAS-Nummer: 3652-89-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR