1-Bromodibenzofuran 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromodibenzo[b,d]furan

OR31661-1G OR31661-250MG OR31661-5G
APOSOR31661-1GEA 29 EUR
APOSOR31661-1G APOSOR31661-250MG APOSOR31661-5G
1-Bromodibenzofuran 99%
1-Bromodibenzofuran
Formel: C₁₂H₇BrO
Molecular Weight: 247,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00094238
CAS-Nummer: 50548-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR