1-Butyl-4-chlorbenzol 96%

Lieferant: Apollo Scientific
Achtung

OR3170-5G OR3170-1G
APOSOR3170-5GEA 92.8 EUR
APOSOR3170-5G APOSOR3170-1G
1-Butyl-4-chlorbenzol 96%
1-Butyl-4-chlorobenzene
Formel: C₁₀H₁₃Cl
Molecular Weight: 168,67 g/mol
Siedepunkt: 225…228 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051926
CAS-Nummer: 15499-27-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR