Methyl-3-oxetane acetate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR317032-1G
OR317032-250MG
APOSOR317032-1GEA
288
EUR
APOSOR317032-1G
APOSOR317032-250MG
Methyl-3-oxetane acetate 97%
Methyl-3-oxetane acetate
Formel:
C₆H₁₀O₃ Molecular Weight: 130.14 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18642557 CAS-Nummer: 1217800-69-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...