2-Methyl-2-propanyl-(3-oxocyclobutyl)carbamat

Lieferant: Apollo Scientific
Achtung

Synonyme: Carbamic acid, N-(3-oxocyclobutyl)-, 1,1-dimethylethyl ester, 2-Methyl-2-propanyl (3-oxocyclobutyl)carbamate, tert-Butyl 3-oxocyclobutylcarbamate

OR317060-1G OR317060-5G
APOSOR317060-1GEA 21.9 EUR
APOSOR317060-1G APOSOR317060-5G
2-Methyl-2-propanyl-(3-oxocyclobutyl)carbamat
tert-Butyl-3-oxocyclobutylcarbamate
Formel: C₉H₁₅NO₃
Molecular Weight: 185,22 g/mol
Schmelzpunkt: 120…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09751872
CAS-Nummer: 154748-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR