3-Aminocyclobutanol 97%

Lieferant: Apollo Scientific
Achtung

OR317063-1G OR317063-250MG OR317063-5G
APOSOR317063-1GEA 377 EUR
APOSOR317063-1G APOSOR317063-250MG APOSOR317063-5G
3-Aminocyclobutanol 97%
3-Aminocyclobutanol
Formel: C₄H₉NO
Molecular Weight: 87,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027510
CAS-Nummer: 4640-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR