(1-Methylcyclobutyl)methanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR317109-100MG OR317109-250MG
APOSOR317109-100MGEA 24.8 EUR
APOSOR317109-100MG APOSOR317109-250MG
(1-Methylcyclobutyl)methanamine hydrochloride
(1-Methylcyclobutyl)methanamine hydrochloride
Formel: C₆H₁₄ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD18074449
CAS-Nummer: 1245647-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR