3-Amino-5-cyclobutyl-1H-pyrazole

Lieferant: Apollo Scientific

OR317113-1G OR317113-5G
APOSOR317113-1GEA 79.9 EUR
APOSOR317113-1G APOSOR317113-5G
3-Amino-5-cyclobutyl-1H-pyrazole
3-Amino-5-cyclobutyl-1H-pyrazole
Formel: C₇H₁₁N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08061045
CAS-Nummer: 326827-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR