Ethyl (3-hydroxycyclobutyl)acetate

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-(3-hydroxycyclobutyl)acetate

OR317120-1G OR317120-250MG
APOSOR317120-1GEA 580 EUR
APOSOR317120-1G APOSOR317120-250MG
Ethyl (3-hydroxycyclobutyl)acetate
Ethyl (3-hydroxycyclobutyl)acetate
Formel: C₈H₁₄O₃
Molecular Weight: 158,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23105932
CAS-Nummer: 1408075-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR