trans-tert-Butyl-3-hydroxycyclobutylcarbamate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl (trans-3-hydroxycyclobutyl)carbamate, trans-1-Amino-3-hydroxycyclobutane, N-BOC protected
OR317138-1G
OR317138-25G
OR317138-5G
APOSOR317138-1GEA
29
EUR
APOSOR317138-1G
APOSOR317138-25G
APOSOR317138-5G
trans-tert-Butyl-3-hydroxycyclobutylcarbamate 97%
trans-tert-Butyl-3-hydroxycyclobutylcarbamate
Formel:
C₉H₁₇NO₃ Molecular Weight: 187,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09038209 CAS-Nummer: 389890-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...