1-Azetidinecarboxylic acid-3-iodophenylmethyl ester

Lieferant: Apollo Scientific

Synonyme: Benzyl 3-iodoazetidine-1-carboxylate, 1-Cbz-3-iodoazetidine

OR317157-1G OR317157-5G
APOSOR317157-1GEA 105 EUR
APOSOR317157-1G APOSOR317157-5G
1-Azetidinecarboxylic acid-3-iodophenylmethyl ester
1-Azetidinecarboxylic acid-3-iodophenylmethyl ester
Formel: C₁₁H₁₂INO₂
Molecular Weight: 317,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09026510
CAS-Nummer: 939759-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR