1-Azetidinecarboxylic acid-3-aminophenylmethyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Aminoazetidine, N1-CBZ-protected, 1-Cbz-3-Aminoazetidine

OR317158-1G OR317158-5G
APOSOR317158-1GEA 78.3 EUR
APOSOR317158-1G APOSOR317158-5G
1-Azetidinecarboxylic acid-3-aminophenylmethyl ester
1-Azetidinecarboxylic acid-3-aminophenylmethyl ester
Formel: C₁₁H₁₄N₂O₂
Molecular Weight: 206,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07772068
CAS-Nummer: 112257-20-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR