Methyl-1-boc-3-azetidineacetate

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 3-(2-methoxy-2-oxoethyl)azetidine-1-carboxylate

OR317161-1G OR317161-25G OR317161-5G
APOSOR317161-1GEA 74.1 EUR
APOSOR317161-1G APOSOR317161-25G APOSOR317161-5G
Methyl-1-boc-3-azetidineacetate
Methyl-1-boc-3-azetidineacetate
Formel: C₁₁H₁₉NO₄
Molecular Weight: 229.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11559001
CAS-Nummer: 497160-14-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR