1-Boc-3-(bromomethyl)azetidine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 3-(bromomethyl)azetidine-1-carboxylate
OR317164-5G
APOSOR317164-5GEA
102
EUR
APOSOR317164-5G
1-Boc-3-(bromomethyl)azetidine
1-Boc-3-(bromomethyl)azetidine
Formel:
C₉H₁₆BrNO₂ Molecular Weight: 250,136 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16556174 CAS-Nummer: 253176-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...